EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Expression of laminin isoforms in mouse myogenic cells in vitro and in vivo.
PMID 8719886
Abstract
The expression of laminin-1 (previously EHS laminin) and laminin-2 (previously merosin) isoforms by myogenic cells was examined in vitro and in vivo. No laminin alpha 2 chainspecific antibodies react with mouse tissues, 50 rat monoclonal antibodies were raised against the mouse laminin alpha 2 chain: their characterization is described here. Myoblasts and myotubes from myogenic cell lines and primary myogenic cultures express laminin beta 1 and gamma 1 chains and form a complex with a 380 kDa alpha chain identified as laminin alpha 2 by immunofluorescence, immunoprecipitation and PCR. PCR from C2C12 myoblasts and myotubes for the laminin alpha 2 chain gene (LamA2) provided cDNA sequences which were used to investigate the in vivo expression of mouse LamA2 mRNA in embryonic tissues by in situ hybridization. Comparisons were made with specific probes for the laminin alpha 1 chain gene (LamA1). LamA2 but not LamA1 mRNA was expressed in myogenic tissues of 14- and 17-day-old mouse embryos, while the laminin alpha 2 polypeptide was localized in adjacent basement membranes in the muscle fibres. In situ hybridization also revealed strong expression of the LamA2 mRNA in the dermis, indicating that laminin alpha 2 is expressed other than by myogenic cells in vivo. Immunofluorescence studies localized laminin alpha 2 in basement membranes of basal keratinocytes and the epithelial cells of hair follicles, providing new insight into basement membrane assembly during embryogenesis. In vitro cell attachment assays revealed that C2C12 and primary myoblasts adhere to laminin-1 and -2 isoforms in a similar manner except that myoblast spreading was significantly faster on laminin-2. Taken together, the data suggest that laminins 1 and 2 play distinct roles in myogenesis.