EMAIL THIS PAGE TO A FRIEND
Ophthalmic research
Interaction of glycyl-phenylalanine with carbohydrates as a model of N-terminal glycation of proteins.
PMID 8727968
Abstract
Glycation is thought to be a factor in the development of cataract in elderly and diabetic patients. In this report, we describe our initial investigations on the reaction of the dipeptide glycyl-phenylalanine (GF) with different carbonyl compounds as a model of protein glycation. The results obtained demonstrate that the interaction of GF with carbohydrates has features in common with glycation of lens membrane proteins and lens crystallins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2752
Gly-Phe
C11H14N2O3