EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Long-range sequence analysis in Xq28: thirteen known and six candidate genes in 219.4 kb of high GC DNA between the RCP/GCP and G6PD loci.
PMID 8733135
Abstract
DNA comprising 219 447 bp was sequenced in nine cosmids and verified at > 99.9% precision. Of the standard repetitive elements, 187 Alus make up 20.6% of the sequence, but there were only 27 MERs (2.9%) and 17 L1 fragments (1.6%). This may be characteristic of such high GC (57%) regions. The sequence also includes an 11.3 kb tract duplicated with 99.2% identity at a distance of 38 kb. The region is 80-90% transcribed and 12.5% translated. Thirteen known genes and their exon-intron borders are all accurately predicted at least in part by GRAIL programs, as are six additional genes. From centromere to telomere, the orientation of transcription varies among the first eight genes, then runs centromeric to telomeric for the next five, and is in the opposite sense for the last six. Eighteen of the 19 genes are associated with CpG islands. Two islands are exact copies in the 11.3 kb repeat units, and could thus give rise to double dosage levels of an X-linked gene. Another island is associated with two genes transcribed in opposite directions. From the sequence data, three genes and their exon structure are inferred. One of them, previously associated with HEX2, is shown to be a different gene unrelated to hexokinases; a second gene, previously known by an EST, is plexin, from its 65.5% identity with the Xenopus analog; and a third is a subunit of a vacuolar H-ATPase, and is named VATPS1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500438
Anti-G6PD (AA 308-320) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA000834
Anti-G6PD antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SRP6505
G6PD human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
G6378 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G7877 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
G4134 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G5885 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, 550-1,100 units/mg protein (biuret)
G8404 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret)
G5760 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
G8529 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, 550-1,100 units/mg protein (biuret)