EMAIL THIS PAGE TO A FRIEND
Biological chemistry Hoppe-Seyler
Synthetic peptides corresponding to alpha-lactalbumin and beta-lactoglobulin sequences with angiotensin-I-converting enzyme inhibitory activity.
PMID 8737991
Abstract
Novel angiotensin-I-converting enzyme (ACE) inhibitory activities were detected in synthetic peptides corresponding to sequences of beta-lactoglobulin and alpha-lactalbumin and which are known to possess opioid activity. Using hippuryl-histidyl-leucine as substrate, the tetrapeptides beta-lactorphin (Tyr-Leu-Leu-Phe), alpha-lactorphin (Tyr-Gly-Leu-Phe) and beta-lactotensin (His-Ile-Arg-Leu) were shown to have IC50 values of 171.8, 733.3 and 1153.2 microM, respectively. Related dipeptides also inhibited ACE, with Tyr-Leu being the most potent, having an IC50 value of 122.1 microM.