EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Synthesis of the very acid-sensitive Fmoc-Cys(Mmt)-OH and its application in solid-phase peptide synthesis.
PMID 8740963
Abstract
S-4-methoxytrityl cysteine was synthesized and converted into the corresponding Fmoc-Cys(Mmt)-OH by its reaction with Fmoc-OSu. As compared to the corresponding Fmoc-Cys(Trt)-OH, the S-Mmt-function was found to be considerably more acid labile. Quantitative S-Mmt-removal occurs selectively in the presence of groups of the tert butyl type and S-Trt by treatment with 0.5-1.0% TFA. The new derivative was successfully utilized in the SPPS of Tyr1-somatostatin on 2-chlorotrityl resin. In this synthesis groups of the Trt-type were exclusively used for amino acid side-chain protection. Quantitative cleavage from the resin and complete deprotection was performed by treatment with 3% TFA in DCM-TES (95:5) for 30 min at RT. We observed no reduction of tryptophan under these conditions.