EMAIL THIS PAGE TO A FRIEND
Current problems in dermatology
Use of penciclovir and famciclovir in the management of genital herpes.
PMID 8743273
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0035
Penciclovir
C10H15N5O3