EMAIL THIS PAGE TO A FRIEND
Wei sheng wu xue bao = Acta microbiologica Sinica
[Studies on bacterial metabolism pathway of 2-naphthoic acid].
PMID 8745549
Abstract
The missing chemical Link (Phthalate) of bacterial metabolism pathway for 2-naphthoic acid has been identified to be existing. The genetic regulation mechanism of 2-NAT strain for 2-naphthoic acid metabolism is also investigated with plasmid curing, transposon Tn10 mutagenesis and mutation reversion experiments. It is discovered that 2-NAT strain carries at least one plasmid. The plasmid and the chromosomal DNA co-encode and control the bacterial metabolism pathway for 2-naphthoic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70400
2-Naphthoic acid, ≥97.0% (GC)
C11H8O2
180246
2-Naphthoic acid, 98%
C11H8O2