EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[18-Hydroxycorticosterone (18-OH-B)].
PMID 8753291
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
710806
18-Hydroxycorticosterone, 97% (CP)
C21H30O5