EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
The multiple-sugar metabolism (msm) gene cluster of Streptococcus mutans is transcribed as a single operon.
PMID 8764489
Abstract
The multiple-sugar metabolism (msm) locus of Streptococcus mutans constitutes a non-PTS sugar uptake system responsible for the transport and utilization of raffinose, melibiose and isomaltotrioses. While previous studies have used polar mutations to suggest that these genes are co-transcribed, there has not been evidence to support this. In this report we present direct evidence that the msm genes can be transcribed as a single operon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0381
Isomaltotriose, ≥98% (TLC)
C18H32O16
47884
Isomaltotriose, DP3, analytical standard
C18H32O16