EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
New low-density lipoprotein receptor upregulators acting via a novel mechanism.
PMID 8765518
Abstract
The synthesis and biological activity of a new series of benzamides and related compounds that upregulate the expression of the low-density lipoprotein (LDL) receptor in human hepatocytes (HepG2 cells) by a novel mechanism are described. The lead compound, N-[5-[(3-cyclohexylpropionyl)amino]-2-methylphenyl]-4-hydroxybe nzamide (1, RPR102359), increased the expression of the LDL receptors in HepG2 cells by 80% when tested at a concentration of 3 microM. Mevinolin (lovastatin) was found to increase the LDL receptor expression by 70% at the same concentration. In contrast to mevinolin, 1 was found to have no effect on cholesterol biosynthesis in liver homogenates or in HepG2 cells at doses where substantial upregulation of the LDL receptor was observed and thus stimulated LDL receptor expression by a novel mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
428949
4-Methyl-3-nitrobenzoyl chloride, 99%
C8H6ClNO3