EMAIL THIS PAGE TO A FRIEND
Der Chirurg; Zeitschrift fur alle Gebiete der operativen Medizen
[Diagnosis and therapy of postoperative chyloperitoneum].
PMID 8767101
Abstract
Chyloperitoneum is a rare complication of surgical procedures. We report a case of severe chyloperitoneum after abdominothoracic esophageal resection with two-field lymphadenectomy. After diagnosis was established, the lymph leak was located with oral iodine-marked fatty acids (123I-pentadecanoic acid) showing increased activation in the right middle abdomen. Because conservative treatment with total parenteral nutrition showed no decrease in chyle volume, relaparatomy was performed and the lymphatic fistula was successfully treated with suture ligatures. Pathophysiology and different options for localization and treatment of postoperative chyloperitoneum are discussed.
Related Materials