EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Early detachment of colon carcinoma cells during CD95(APO-1/Fas)-mediated apoptosis. I. De-adhesion from hyaluronate by shedding of CD44.
PMID 8769429
Abstract
Ligation of CD95 (APO-1/Fas) cell surface receptors induces death in apoptosis-sensitive cells. Induction of apoptosis in adherent gamma interferon-stimulated HT-29 and COLO 205 colon carcinoma cells by cross-linking CD95 with anti-APO-1 monoclonal antibody resulted in detachment of the cells from hyaluronate starting about 1 h after antibody exposure. Loss of adhesion was paralleled by a substantial reduction of the multifunctional cell surface adhesion molecule CD44. As evidenced by cycloheximide treatment, this effect was not caused by impaired protein synthesis. Depletion of surface CD44 was also not due to membrane blebbing, since cytochalasin B failed to inhibit ascension from hyaluronate. Instead, ELISA and time kinetics showed increasing amounts of soluble CD44 in the supernatant of CD95-triggered cells. SDS-PAGE revealed that soluble CD44 had an apparent molecular mass of about 20 kD less than CD44 immunoprecipitated from intact cells. Thus, CD95-triggering induced shedding of CD44. Shedding is a novel mechanism operative in early steps of CD95-mediated apoptosis. Shedding surface molecules like CD44 might contribute to the active disintegration of dying epithelial cells in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9150
N-Lauroylsarcosine sodium salt, detergent for use in cell lysis
C15H28NNaO3