EMAIL THIS PAGE TO A FRIEND
The Surgical clinics of North America
The critical care of the severely injured patient--I. Assessing and improving oxygen delivery.
PMID 8782482
Abstract
The hallmark of critical care of the major trauma victim is the assurance of adequate cellular perfusion of oxygen and nutrients. Although controversy exists as to the exact endpoint of therapeutic maneuvers aimed at this assurance, there should be general agreement that benefit is to be gained from careful observation of clinical signs of perfusion status and bedside determinations of oxygen transport parameters.