EMAIL THIS PAGE TO A FRIEND
Neuron
The sweet, sweet smell of diacetyl.
PMID 8785043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11038
2,3-Butanedione, analytical standard
C4H6O2