EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Offset printer's occupational allergic contact dermatitis caused by cobalt-2-ethylhexoate.
PMID 8789236
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
444545
Cobalt(II) 2-ethylhexanoate solution, 65 wt. % in mineral spirits
C16H30CoO4