EMAIL THIS PAGE TO A FRIEND
Japanese journal of pharmacology
Potentiation of L-dopa-induced behavioral excitement by histamine H1-receptor antagonists in mice.
PMID 8791174
Abstract
Effects of histamine H1-receptor antagonists on L-dopa-induced behavioral excitement were examined in mice to confirm behaviorally the inhibition of dopamine uptake by these compounds. L-Dopa (100-300 mg/kg, s.c.) combined with pargyline hydrochloride (80 mg/kg, i.p.) caused a dose-dependent behavioral excitement. The marked excitement induced by L-dopa (300 mg/kg) plus pargyline was significantly inhibited by pimozide (0.1 - 1 mg/kg, s.c.), a selective dopamine antagonist. Tripelennamine (10 mg/kg, s.c.), d-chlorpheniramine (1 and 2 mg/kg, s.c.), homochlorcyclizine (2 and 5 mg/kg, s.c.), diphenhydramine (2 and 5 mg/kg, s.c.) and mepyramine (2 and 5 mg/kg, s.c.) each markedly enhanced the moderate behavioral excitement induced by L-dopa (150 mg/kg) plus pargyline. These findings are behavioral evidence for inhibition of dopamine uptake by H1 antagonists, which has been suggested by neurochemical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7511
Tripelennamine hydrochloride, analytical standard
C16H21N3 · HCl