EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
In vivo and in vitro measurement of CYP2C19 activity.
PMID 8791767
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC126
4-Hydroxymephenytoin
C12H14N2O3
H146
4-Hydroxymephenytoin, ≥98% (HPLC)
C12H14N2O3