EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Expression and characterization of human pancreatic preprocarboxypeptidase A1 and preprocarboxypeptidase A2.
PMID 8806703
Abstract
We are investigating the potential utility of human carboxypeptidases A in antibody-directed enzyme prodrug therapy (ADEPT). Hybridization screening of a human pancreatic cDNA library with cDNA probes that encoded either rat carboxypeptidase A1 (rCPA1) or carboxypeptidase A2 (rCPA2) was used to clone the human prepro-CPA homologs. After expression of the respective pro-hCPA cDNA in Saccharomyces cerevisiae, the enzymes were purified to homogeneity by a combination of hydrophobic and ion-exchange chromatography. Purified hCPA1 and hCPA2 migrate as a single protein band with M(r) 34,000 when subjected to gel electrophoresis in the presence of sodium dodecyl sulfate under reducing conditions. Kinetic studies of the purified enzymes with hippuryl-L-phenylalanine resulted in kcat/Km values of 57,000 and 19,000 M-1 s-1 for hCPA1 and hCPA2, respectively. Using the ester substrate, hippuryl-D, L-phenyllactate, we found unique esterase/ peptidase specific activity ratios among hCPA1, hCPA2, rCPA1, and bovine CPA (bCPA) ranging from 13 to 325. Two potential ADEPT substrates, methotrexate-alpha-phenylalanine (MTX-Phe) and methotrexate-alpha-(1-naphthyl)alanine (MTX-naphthylAla) were also analyzed. The kcat/Km values for MTX-Phe were 440,000 and 90,000 M-1 s-1 for hCPA1 and hCPA2, respectively, and for MTX-naphthylAla these values were 1400 and 1,400,000 M-1 s-1 for hCPA1 and hCPA2, respectively. The kinetic data show that hCPA2 has a larger substrate binding site than the hCPA1 enzyme. Differences between hCPA1 and hCPA2 were also observed in thermal stability experiments at 60 degrees C where the half-life for thermal denaturation of hCPA2 is eightfold longer than that for hCPA1. These experiments indicate that hCPA1 and hCPA2 are potential candidates for use in a human-based ADEPT approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6875
Hippuryl-L-phenylalanine
C18H18N2O4