EMAIL THIS PAGE TO A FRIEND
The Journal of eukaryotic microbiology
In vitro evaluation of anticryptosporidial agents using MDCK cell culture and chemiluminescence immunoassay.
PMID 8822878
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
MDCK Cell Line canine