EMAIL THIS PAGE TO A FRIEND
The Journal of eukaryotic microbiology
Gram-chromotrope: a new technique that enhances detection of microsporidial spores in clinical samples.
PMID 8822884
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3143
Chromotrope 2R, suitable for modified Gomori Trichrome stain
C16H10N2Na2O8S2