EMAIL THIS PAGE TO A FRIEND
Pancreas
Kinetic determination of amylase on microtiter plates: an improved substrate.
PMID 8829193
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93834
2-Chloro-4-nitrophenyl-α-D-maltotrioside, ≥96.0% (HPLC)
C24H34ClNO18