EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of the pharmaceutical peptide TP9201 by post-column reaction with copper(II) followed by electrochemical detection.
PMID 8833978
Abstract
An electrochemical detection method was applied to the determination of the synthetic peptide TP9201 (Telios Pharmaceuticals). The method utilizes reversed-phase HPLC, followed by post-column formation of Cu(II)-peptide complexes to render peptides electrochemically active via the Cu(III/II) couple. TP9201 is cyclic and N-amidated; the lack of a free amine precludes the use of typical fluorescent labeling reagents. Neither the cyclic structure nor the N-amidation prevented the copper complexation reaction, however. The detection limit in bovine serum was 20 nM, limited by interfering sample peaks, and the detector response was linear in a range 10-400 nM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15270
Biuret, 98%
C2H5N3O2