EMAIL THIS PAGE TO A FRIEND
Journal of bioluminescence and chemiluminescence
Super-sensitive enzyme immunoassay for thyroid stimulating hormone using a new synergistic enhanced chemiluminescent endpoint.
PMID 8844344
Abstract
The enhancers 1,1'-biphenyl-4-yl boronic acid and 4-iodophenol act synergistically in the horseradish peroxidase-catalysed oxidation of luminol. This concentration-dependent effect reduces background, increases signal and hence improves signal/background for detection of peroxidase. The same type of synergistic effect was found when 1,1'-biphenyl-4-yl boronic acid was added to a commercial enhanced chemiluminescence signal reagent (Amerlite Signal Reagent). This synergistic enhanced chemiluminescent endpoint (Amerlite Signal Reagent containing 1,1'-biphenyl-4-yl boronic acid) for a horseradish peroxidase label has been tested in the Amerlite TSH and the Amerlite TSH-30 Ultrasensitive assays. The detection limit (mean of 20 replicates of the zero standard+2SD) in the Amerlite TSH assay was 0.0029 mIU/L, and in the Amerlite TSH-30 Ultrasensitive assay the detection limit was 0.0005 mIU/L using the synergistic enhanced endpoint. Reassessment of the detection limit using a 1:40 dilution of the first standard (0.119 mIU/L) as the lowest assay standard gave a value of 0.0015 mIU/L for the Amerlite TSH-30 Ultrasensitive assay with the synergistic endpoint. A limited (n = 29) method comparison using samples from euthyroid, hyperthyroid and hypothyroid patients revealed excellent correlation between the conventional and synergistic TSH immunoassays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
483451
4-Biphenylboronic acid, ≥95.0%
C12H11BO2
I10201
4-Iodophenol, 99%
C6H5IO