EMAIL THIS PAGE TO A FRIEND
Folia microbiologica
Metabolism of 2-chlorobenzoic acid in Pseudomonas stutzeri.
PMID 8846991
Abstract
3-Chloropyrocatechol is formed as a result of oxidation of 2-chlorobenzoate by Pseudomonas stutzeri. 2-Chloro-cis,cis-muconic acid is the product of oxidation of 3-chloropyrocatechol. A catabolic pathway for the degradation of 2-chlorobenzoate by a newly isolated strain of P. stutzeri is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135577
2-Chlorobenzoic acid, 98%
C7H5ClO2
506877
2-Chlorobenzoic acid, analytical standard
C7H5ClO2