EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous quantitation of buspirone and its major metabolite 1-(2-pyrimidinyl)piperazine in human plasma by high-performance liquid chromatography with coulometric detection.
PMID 8851759
Abstract
Buspirone is a member of the azapirone group of anxiolytic drugs and has one major metabolite, 1-(2-pyrimidinyl)piperazine (1-PP). The analyte, its metabolite and the internal standard were extracted from plasma utilizing solid-phase extraction columns. Chromatography was performed using isocratic reversed-phase high-performance liquid chromatography with coulometric end-point detection. The calibration graph was linear over the range 0-50 ng ml-1 of plasma. The lower limits of quantitation for buspirone and 1-PP were 0.5 and 2 ng ml-1, respectively, when 1 ml of plasma was extracted. The intra-assay relative standard deviations (RSD) over the range of the calibration graph varied from 4 to 12.5% for buspirone and 1-PP. The inter-assay RSD was 6.9% for 1-PP and 9.6% for buspirone. The recovery averaged 96% for buspirone and 66% for 1-PP. Plasma profiles of buspirone and 1-PP following oral dosing are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
421235
1-(2-Pyrimidyl)piperazine, 98%
C8H12N4