EMAIL THIS PAGE TO A FRIEND
Journal of applied toxicology : JAT
Sulfur dioxide.
PMID 8854224
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295698
Sulfur dioxide, ≥99.9%
O2S