EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
GC-MS profiling of urinary organic acids evaluated as a quantitative method.
PMID 8855143
Abstract
We assessed the quantitative performances of a classical method for profiling urinary organic acids: ethyl acetate extraction/oxime-trimethylsilyl derivatization/GC-MS. Twenty-seven acids were quantified on the basis of specific ions in both scan and selected-ion monitoring modes. We found that the tuning of the mass detector severely affects the calibration factors, being critical to achieve quantitative results, and we propose a practical procedure for reproducible tuning. Of seven compounds tested, tropic acid was retained as the internal standard suitable for most of the acids of clinical interest; a second internal standard, 2-ketocaproic acid, was used in quantifying keto-acids. The within-day and total relative standard deviations (CVs), estimated from scan-mode analyses of urine, ranged from 2.6% to 12.7% and from 4.2% to 11.8%, respectively. Curvilinear relationships between analytical response and concentration were observed for most of the acids investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6625
2-Ketohexanoic acid sodium salt, 97.0-103.0%
C6H9NaO3
T89206
Tropic acid, 98%
C9H10O3