EMAIL THIS PAGE TO A FRIEND
Journal of the National Medical Association
Hyperpigmentation associated with selenium sulfide lotion.
PMID 8855644
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se