EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
The Lck tyrosine kinase is expressed in brain neurons.
PMID 8858916
Abstract
The lck gene product, p56lck, is a member of the src-related family of protein tyrosine kinases. It is known as lymphocyte specific and involved in thymocyte development and in the immune response mediated by the T cell receptor. We report that the lck gene is also expressed in adult mouse CNS and that brain p56lck is similar to the thymus protein. In situ hybridization and immunohistochemistry show that the lck gene is expressed in neurons throughout the brain in distinct regions, including hippocampus and cerebellum. In primary cultures from fetal mouse brain, neuronal cells are immunoreactive to Lck antiserum. This suggests that the lck gene product might be involved in a new signal transduction pathway in mouse brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-435
Anti-Lck Antibody, clone 3A5, clone 3A5, Upstate®, from mouse