EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Potent mutagenic potential of 4-methylquinoline: metabolic and mechanistic considerations.
PMID 8860954
Abstract
4-Methylquinoline (4-MeQ) showed an extraordinarily potent mutagenicity when compared to quinoline and isomeric methylquinolines. The major metabolite of 4-MeQ was 4-hydroxymethylquinoline, which was not mutagenic under the assay condition employed. Deuteration of the methyl group of 4-MeQ resulted in a decrease in the amount of the hydroxymethyl metabolite and an increase in mutagenicity, indicating that hydroxylation of the substituent methyl group is a detoxication process. A 3-chloro derivative of 4-MeQ was proven to be non-mutagenic. 4-Ethyl-quinoline, as well as 4-hydroxymethylquinoline, was much less mutagenic than 4-MeQ. Taking account of the structure-mutagenicity relationship, a possible mechanism is proposed for the potent mutagenic potential of 4-MeQ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W513903
4-Methylquinoline, ≥99%
C10H9N
158283
Lepidine, 99%
C10H9N