EMAIL THIS PAGE TO A FRIEND
Journal of smooth muscle research = Nihon Heikatsukin Gakkai kikanshi
[Mechanism of increased gastric motility effect of EM523].
PMID 8867984
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05238
Erythromycin A enol ether, analytical standard
C37H65NO12