EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Metabolism of tetramethrin isomers in rat. III. Stereochemistry of reduced metabolites.
PMID 8868003
Abstract
1. Three main urinary metabolites, two isomers of 3-hydroxycyclohexane-1,2-dicarboximide (3-OH-HPI-1 and 2) and 1,2-tetrahydrodicarboxylic acid (TCDA) were purified from rat treated with (1RS, trans)-tetramethrin [3,4,5,6-tetrahydrophthalimidomethyl (1RS, trans)-chrysanthemate]. 2. To elucidate the mechanism of formation of these reduced metabolites, the stereochemistry of 3-OH-HPI-1, 3-OH-HPI-2 and TCDA was clarified by chemical reactions, spectroanalysis (nmr) and X-ray analysis. 3. The sole difference in configuration between 3-OH-HPI-1 and 3-OH-HPI-2 was found to be the orientation of the hydroxyl group to the cyclohexane ring, and both of these reduced metabolites showed cis-addition of two hydrogens. In contrast, reduction resulted in the trans form with TCDA. 4. These findings indicate the existence of two different reduction reaction mechanisms in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45681
Tetramethrin, PESTANAL®, analytical standard
C19H25NO4