EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
The involvement of cytochrome P450 peroxidase in the metabolic bioactivation of cumene hydroperoxide by isolated rat hepatocytes.
PMID 8870978
Abstract
Organic hydroperoxides are believed to be primarily detoxified in cells by the GSH peroxidase/GSSG reductase system and activated to cytotoxic radical species by non-heme iron. However, organic hydroperoxides seem to be bioactivated by cytochrome P450 (P450) in isolated hepatocytes as various P450 (particularly P450 2E1) inhibitors inhibited cumene hydroperoxide (CumOOH) metabolism and attenuated subsequent cytotoxic effects including antimycin A-resistant respiration, lipid peroxidation, iron mobilization, ATP depletion, and cell membrane disruption. CumOOH metabolism was also faster in P450 1A-induced hepatocytes and was inhibited by the P450 1A inhibitor alpha-naphthoflavone. The ferric chelator deferoxamine also prevented cytotoxicity even after CumOOH had been metabolized but had no effect on CumOOH metabolism. This emphasizes the toxicological significance of the iron released following hydroperoxide metabolic activation by cytochrome P450. The radical trap, 4-hydroxy-2,2,6,6-tetramethylpiperidine-N-oxyl (TEMPO), had no effect on CumOOH metabolism but prevented CumOOH-induced antimycin A-resistant respiration, lipid peroxidation, iron mobilization, and loss of membrane integrity. These results suggest that CumOOH is metabolically activated by some P450 enzymes (e.g., P450 2E1) in hepatocytes to form reactive radical metabolites or oxidants that cause lipid peroxidation and cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357774
1-Phenylimidazole, 97%
C9H8N2