EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Hand involvement in allergic contact dermatitis from mercaptobenzothiazole in shoes.
PMID 8879935
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D218154
2,2′-Dithiobis(benzothiazole), 99%
C14H8N2S4