EMAIL THIS PAGE TO A FRIEND
Pharmacology & toxicology
Effects of carbazochrome sodium sulfonate (AC-17) on oleic acid-induced lung injury.
PMID 8884890
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5877
Adrenochrome semicarbazone
C10H12N4O3