EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Cyclic AMP-induced differentiation increases the synthesis of extracellular superoxide dismutase in rat C6 glioma.
PMID 8886798
Abstract
The effect of membrane permeable cAMP analogues on the expression of extracellular superoxide dismutase (EC-SOD) was studied in rat C6 glioma. EC-SOD is constitutively expressed but stimulation with cAMP analogues still increased the EC-SOD transcription and the secreted SOD activity. The potency to enhance EC-SOD expression is correlated with the ability of the cAMP analogue to induce cAMP-dependent differentiation in C6. The increase in EC-SOD mRNA and in secreted activity depended on the concentration of the cAMP analogues and on the cultivation time. Twenty-four hours after addition of 0.5 mM N6, O'2-dibutyryl cAMP (dbcAMP) or N6-monobutyryl cAMP (N6-mbcAMP) EC-SOD mRNA expression increased approximately twofold, while stimulation for 68 h with 0.5 mM N6-mbcAMP or 1 mM 8-Chloro cAMP (ClcAMP) and 1 mM dbcAMP enhanced the mean secreted activity/cell three- and fivefold, respectively. O'2-monobutyryl cAMP (O'2-mbcAMP) did not affect EC-SOD synthesis. The enhancement in EC-SOD activity did not require activation of protein kinase A. ATP, TGF-beta, IFN-gamma, and LPS did not affect EC-SOD synthesis. The presented data point to a cAMP-dependent pathway for the enhanced expression of EC-SOD by glial cells in brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1380
N6-Monobutyryladenosine 3′:5′-cyclic monophosphate sodium salt
C14H17N5NaO7P