EMAIL THIS PAGE TO A FRIEND
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme
Effect of CL316,243, a highly specific beta 3-adrenoceptor agonist, on lipolysis of human and rat adipocytes.
PMID 8886827
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5976
CL 316,243 hydrate, ≥98% (HPLC), powder
C20H18ClNNa2O7 · xH2O