EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Automated analytical systems for drug development studies. V. A system for enzyme kinetic studies.
PMID 8887716
Abstract
Two similar automated analytical systems using liquid chromatography (LC) and microdialysis as an on-line sampling technique were applied to studies of enzyme kinetics. 2',3',5'-Triacetyl-6-azauridine (azaribine) with porcine liver esterase (PLE) and N-acetylphenylalanyl-3,5-diiodotyrosine (AcFY') with pepsin were used as model compounds. The microdialysis sampling technique permitted the rapid separation of low molecular weight analytes from macromolecules, thus simultaneously achieving clean-up of the samples and quenching of the reaction. The combination of rapid LC analysis and microdialysis sampling provided selectivity and automation. The systems are rugged and give reproducible results in agreement with those from manual sampling methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1882
6-Azauridine
C8H11N3O6