EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Pesticide residues in artichokes: effect of different head shape.
PMID 8896357
Abstract
Residues of three pesticides (dimethoate, parathion, and pyrazophos) in two artichoke cultivars, Masedu and Spinoso sardo, were investigated. The amount of pesticides in artichokes was greatly affected by the head shape. In the case of the calix-shaped Masedu artichoke, the residues in whole heads at commercial ripening were on average about twice higher than those of the pagoda-shaped Spinoso sardo artichoke. In the heart this ratio was 4 to 42 times greater. Residue decay rates were very fast, mainly owing to the dilution effect due to head growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45648
Pyrazophos, PESTANAL®, analytical standard
C14H20N3O5PS