EMAIL THIS PAGE TO A FRIEND
Brain research
Lack of development of tolerance to anticonvulsant effects of two excitatory amino acid antagonists, CGP [corrected] 37849 and CGP 39551 in genetically epilepsy-prone rats.
PMID 8896813
Abstract
Two selective excitatory amino acid antagonists, DL-(E)-2-amino-4-methyl- 5-phosphono-3-pentenoic acid (CGP 37849) and its carboxyethylester (CGP 39551), were studied against audiogenic seizures in genetically epilepsy-prone rats following oral administration. Acute administration of CGP 37849 attenuated the clonic and tonic phases of the audiogenic seizures (109 dB, 12-16 kHz) 120 min after pretreatment (ED50 19.7 and 11.2 mumol kg-1, respectively). Similarly, CGP 39551 attenuated the clonic and tonic phases of audiogenic seizures 120 min after acute treatment with ED50 values of 17.2 and 8.8 mumol kg-1, respectively. For chronic studies animals were treated orally once daily (at 10 h) for 4 weeks with CGP 37849 (20 or 40 mumol kg-1) or CGP 39551 (15 or 30 mumol kg-1). In order to assess anticonvulsant activity, rats were subjected to auditory stimulation 120 min after drug administration on days 1, 3 and 5 and then every 3 or 4 days. Following 2 and 4 weeks of repeated drug administration with CGP 37849 (20 and 40 mumol kg-1) the ED50 values against clonic and tonic seizures were not significantly different from those observed following an acute administration. Similarly, 2 and 4 weeks after repeated treatment CGP 39551 (15 and 30 mumol kg-1) the ED50 values against clonic and tonic seizures were not significantly different from those observed following an acute administration. There was no significant difference between the ED50 values following either acute or repeated treatment of the two excitatory amino acid antagonists suggesting a lack of development tolerance. The duration of anticonvulsant activity observed between 0.5 and 24 h following administration of CGP 37849- and CGP 39551 was similar in acute and chronic treatment. The effects of CGP 37849 and CGP 39551 on motor behaviour was also evaluated following acute and repeated treatment by a rotarod apparatus 110 min following drug administration. The TD50 values for CGP 37849 and CGP 39551-induced impairment of locomotor performance recorded 2 or 4 weeks of repeated administration were not significantly different from those observed following an acute administration. The TD50 values for CGP 37849- and CGP 39551-induced impairment of locomotor performance were 87.6 and 70.8 mumol kg-1 i.p. respectively following 2 weeks treatment and 92.9 and 76.9 mumol kg-1 i.p. respectively following 4 weeks treatment. The doses of CGP 37849 and CGP 39551 required to elicit motor impairment were at least an order of magnitude above required for anticonvulsant activity. Since these compounds showed anticonvulsant properties after oral administration and lack of development of tolerance after repeated treatment, a potential use for antiepileptic therapy in man is suggested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0058
CGP 39551, ≥98% (HPLC)
C8H16NO5P