EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Activator-dependent transcription by mammalian RNA polymerase II: in vitro reconstitution with general transcription factors and cofactors.
PMID 8902796