EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
A comparative study of tissue distribution and excretion among three substances implicated in eosinophilia-myalgia syndrome.
PMID 8906290
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0500
1,2,3,4-Tetrahydroharmane-3-carboxylic acid
C13H14N2O2