EMAIL THIS PAGE TO A FRIEND
Journal of pineal research
CGP 52608-induced cyst formation in dinoflagellates: possible involvement of a nuclear receptor for melatonin.
PMID 8912235
Abstract
Melatonin has been shown to regulate gene transcription through RZR/ROR nuclear receptors in mammalian cells. Thiazolidine dione CGP 52608 is a selective agonist of RZR/ROR receptors with little or no affinity for the cell surface G protein-coupled melatonin receptors. In this study, we addressed whether nuclear signaling may be involved in indoleamine-induced encystment of the unicellular dinoflagellates by examining their responses to CGP 52608. Three species of dinoflagellates (Alexandrium catenella, Amphidinium carterae, and Crypthecodinium cohnii) encysted in the presence of CGP 52608 and the responses were reversible and dose-dependent. Since a previous study has implicated the involvement of G proteins in mediating indoleamine-induced encystment of dinoflagellates, we explored the possibility of cross-talks between G protein-dependent and nuclear signaling pathways. The responses of A. catenella to either mastoparan (a direct activator of mammalian G proteins) or indoleamines were assessed in the presence or absence of CGP 52608. Interestingly, CGP 52608 synergized with either indoleamines or mastoparan to produce a more rapid encystment response. These findings suggest that nuclear signaling may be involved in the indoleamine-induced encystment of dinoflagellates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5749
CGP 52608, ≥98%, solid
C8H12N4OS2