EMAIL THIS PAGE TO A FRIEND
The Plant cell
Host-selective toxins: agents of compatibility.
PMID 8914323
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7270
HC toxin from Helminthosporium carbonum, lyophilized powder
C21H32N4O6