EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Speciation of arsenic oxides using laser desorption/ionization time-of-flight mass spectrometry.
PMID 8916457
Abstract
Positive and negative ion mass spectra of arsenic trioxide (As2O3) and arsenic pentaoxide (As2O5) have been obtained by single-step laser desorption/ionization time-of-flight mass spectrometry. Pulsed UV radiation at 266 nm was used for the simultaneous desorption and ionization of the solid sample. High-mass cluster ions that are unique to the oxidation state of each oxide sample appear in the negative ion mass spectra. The As2O3 produces As3O5-, while the As2O5 yields As3O8-. The formation of unique negative cluster ions presents the capability for arsenic oxidation state speciation by laser desorption/ionization mass spectrometry. The ability of time-of-flight mass spectrometry to examine the relative amounts of each arsenic oxide present in a series of mixtures is discussed. Application of our speciation technique to a model incinerator sample is demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
483257
Arsenic(V) oxide, 99%
As2O5