EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
The E5 gene product of rhesus papillomavirus is an activator of endogenous Ras and phosphatidylinositol-3'-kinase in NIH 3T3 cells.
PMID 8917513