EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Methodical analysis of protein-nitrocellulose interactions to design a refined digestion protocol.
PMID 8921181
Abstract
We have analyzed the efficacies of seven different organic solvents, 13 organic bases, and 17 detergents to dissociate electroblotted proteins from nitrocellulose. Most efficient were a 1% piperidine-40% acetonitrile mixture and 1% concentrations of either cetyltrimethylammonium bromide or several polyoxyethylene and zwitterionic detergents at pH > or = 8.5. In general, detergent-promoted elution varied with pH (8.5-->12.0) and temperature (37-->65 degrees C) in a detergent-dependent and protein-dependent manner, making for unpredictable results of any combination. However, Zwittergent 3-16, as the major exception, eluted proteins of different sizes and properties almost equally well at pH 8.5. This preferred effect is not enhanced by addition of organic solvent. In fact, detergent/acetonitrile mixtures were generally less efficient for protein elution. Zwittergent 3-16 (1% in 100 mM NH4HCO3) proved to be the most powerful additive for one-step enzymatic digestion of nitrocellulose-bound proteins, both in terms of highest recoveries (including endoproteinase Lys-C digestions) and general applicability, a major improvement over the use of Tween 80 and hydrogenated Triton X-100. The new digest procedure takes just 2 h to complete and requires 5 ng protease per 1 microliter buffer per 1 mm2 of nitrocellulose; relatively large pieces of membrane (> or = 100 mm2) can be successfully processed. The same detergent is a satisfactory additive for digestion of polyvinylidene difluoridebound proteins as well. Moreover, Zwittergent 3-16 is fully compatible with the usual downstream procedures. It is uv (OD214) transparent and does not interfere with high-performance liquid chromatography in any other way. Apparent incompatibility with direct (i.e., "prechromatography") matrix-assisted laser-desorption ionization time-of-flight mass spectrometry can be either reduced by methanol washing of the dried sample and activation of a low-mass gate or eliminated by a simple microliter cleanup procedure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6883
3-(N,N-Dimethylpalmitylammonio)propanesulfonate, ≥98% (TLC)
C21H45NO3S