EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Prediction of the rodent carcinogenicity of organic compounds from their chemical structures using the FALS method.
PMID 8933054
Abstract
Fuzzy adaptive least-squares (FALS), a pattern recognition method recently developed in our laboratory for correlating structure with activity rating, was used to generate quantitative structure-activity relationship (QSAR) models on the carcinogenicity of organic compounds of several chemical classes. Using the predictive models obtained from the chemical class-based FALS QSAR approach, the rodent carcinogenicity or noncarcinogenicity of a group of organic chemicals currently being tested by the U.S. National Toxicology Program was estimated from their chemical structures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462268
4,4′-Diamino-2,2′-stilbenedisulfonic acid, technical grade, 85%
C14H14N2O6S2