EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Adhesion among neural cells of the chick embryo. II. Purification and characterization of a cell adhesion molecule from neural retina
PMID 893446