EMAIL THIS PAGE TO A FRIEND
Anesthesia progress
Premedication with chlordemethyldiazepam and anxiolytic effect of diazepeam in implantology.
PMID 8934975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7662
Delorazepam
C15H10Cl2N2O